Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445530010
1 g, Each
Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445530010
1 g, Each
Quantity
Catalog number: 445530010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1142363-56-1
IUPAC Name
4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3,6-dihydro-2H-pyran-4-yl)-1,3,2-dioxaborolane
Molecular Formula
C15H27BO3
InChI Key
VOXXPMAJNSVUKJ-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC(C)(C)OC(C)(C)C1
Appearance (Color)
White to off-white
Melting point
91°C to 96°C
GC
>=96.0 %
NMR (1H-NMR)
Consistent with structure
Appearance (Form)
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text