3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals
3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445560010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name[3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Molecular FormulaC7H7BN4O2
InChI KeyXMJKBDSKRITXBW-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC=C1)C1=NNN=N1
Molecular Weight (g/mol)189.97
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Appearance (Form)Solid
HPLC>=95.0 %
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only