4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific Chemicals
4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445570010
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
140.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
140.00 
Save 20.35 (15%)
Add to cart
4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific Chemicals
Catalog number445570010
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
140.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS179942-55-3
IUPAC Name[4-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Molecular FormulaC7H7BN4O2
InChI KeyDXUPJOQUAAVAGV-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1=NNN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
HPLC>=95.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only