Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
Glass Bottle
Catalog number 445660025
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
Add to cart
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Catalog number445660025
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 390 nm A: =<0.5
UVat 410 nm A: =<0.05
Appearance (Form)Clear liquid
GC>=99.85 %
Residue after evaporation=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only