Tetrachloroethylene, 99+%, for HPLC, Thermo Scientific Chemicals
Tetrachloroethylene, 99+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrachloroethylene, 99+%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
Glass bottle
Catalog number 445690025
Price (USD)/ Each
280.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
330.00 
Save 49.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
280.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
330.00 
Save 49.35 (15%)
Add to cart
Tetrachloroethylene, 99+%, for HPLC, Thermo Scientific Chemicals
Catalog number445690025
Price (USD)/ Each
280.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
330.00 
Save 49.35 (15%)
-
Add to cart
Chemical Identifiers
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Residue after evaporation=<0.0005 %
UVat 295 nm A: =<0.3
UVat 400 nm A: =<0.03
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only