1-Propanol, ACS reagent
1-Propanol, ACS reagent
1-Propanol, ACS reagent
1-Propanol, ACS reagent
Thermo Scientific Chemicals

1-Propanol, ACS reagent

CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 445770010
Price (USD)
76.65
Special offer
Online exclusive
89.80
Save 13.15 (15%)
Each
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
89.80
Save 13.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Water=<0.2 %
Residue after evaporation=<0.001 %
Carbonyl compounds=<0.03 % (as C2H5CHO)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link