Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 445790010
1 L, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 445790010
1 L, Each
Quantity
Catalog number: 445790010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Boron (B)
=<1 ppm
Chromium (Cr)
=<1 ppm
Iron (Fe)
=<1 ppm
Appearance (Form)
Clear liquid
Antimony (Sb)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text