1-Methyl-2-pyrrolidinone, 99.5%, for analysis
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for analysis

CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 445790025
Price (USD)
250.65
Special offer
Online exclusive
Ends: 26-Sep-2025
295.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
250.65
Special offer
Online exclusive
Ends: 26-Sep-2025
295.00
Save 44.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Boron (B)=<1 ppm
Chromium (Cr)=<1 ppm
Iron (Fe)=<1 ppm
Appearance (Form)Clear liquid
Antimony (Sb)=<1 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link