Applied Biosystems™

OpenArray™ AccuFill™ System Tips

Catalog number: 4458107
Have Questions?
Applied Biosystems™

OpenArray™ AccuFill™ System Tips

Catalog number: 4458107
Have Questions?
Catalog Number
4458107
Unit Size
10 boxes
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 box
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
445810710 boxes
-
44572461 box
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

This is a box of tips for use with the OpenArray™ AccuFill™ System, which is part of the OpenArray™ Real-Time PCR Platform. Since AccuFill tips have a shorter shelf-life than OpenArray plates, we recommend that you align your order quantity with your estimated OpenArray throughput.

About the OpenArray Real-Time PCR Platform
The OpenArray Real-Time PCR Platform provides PCR-based genotyping and gene expression analysis of hundreds to thousands of samples and assays per day. Researchers can analyze up to 144 samples on a single OpenArray plate. Three OpenArray plates can be cycled and imaged simultaneously on the OpenArray™ Real-Time PCR Instrument, providing high-throughput results with ease. The system workflow thermal cycles and fluorescently images the OpenArray plates. Results are captured on the external PC, and the OpenArray analysis software enables researchers to view specific assay data.

Complete Platform
The OpenArray Real-Time PCR Platform includes:
• OpenArray Real-Time PCR Instrument—images three OpenArray plates simultaneously
• OpenArray AccuFill System—loads samples into OpenArray plates
• OpenArray™ Case Sealing Station—seals OpenArray plates
• OpenArray instrument accessories (includes PC system)
• OpenArray™ Genotyping Analysis Software
• OpenArray™ Real-Time PCR Analysis Software

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Tips
For Use With (Equipment)
OpenArray™
Product Line
AccuFill™, OpenArray™
Product Type
System Tip
For Use With (Application)
Real Time PCR (qPCR)
Quantity
10 Boxes

Contents & Storage

Each Pk. contains 10 Boxes of AccuFill™ tips, with 384 tips / box.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text