Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 445831000
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
Add to cart
Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Catalog number445831000
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
Molecular Weight (g/mol)112.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to amber
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Liquid
Strength1.90 to 2.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only