Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Thermo Scientific Chemicals

Hydrogen chloride, 3M solution in CPME, AcroSeal™

CAS: 7647-01-0 | ClH | 36.46 g/mol
Catalog NumberQuantityPackaging
445851000100 mLAcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445851000
Price (JPY)
-
Request A Quote
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Strength2.85 to 3.30 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link