Thermo Scientific Chemicals

2-Bromoethyl ether, 90%, Thermo Scientific Chemicals

Catalog number: 445860250
25 mL, Each
Thermo Scientific Chemicals

2-Bromoethyl ether, 90%, Thermo Scientific Chemicals

Catalog number: 445860250
25 mL, Each
Quantity
Catalog number: 445860250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5414-19-7
IUPAC Name
1-bromo-2-(2-bromoethoxy)ethane
Molecular Formula
C4H8Br2O
InChI Key
FOZVXADQAHVUSV-UHFFFAOYSA-N
SMILES
BrCCOCCBr
Appearance (Color)
Colorless to brown
Appearance (Form)
Liquid
Appearance
May darken on storage
Infrared spectrum
Conforms
GC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text