Applied Biosystems™

BigDye™ Direct Cycle Sequencing Kit

Catalog number: 4458689
Have Questions?
Applied Biosystems™

BigDye™ Direct Cycle Sequencing Kit

Catalog number: 4458689
Have Questions?
Catalog Number
4458689
Unit Size
24 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
445868924 reactions
-
44586881,000 reactions
-
4458687100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The BigDye™ Direct Cycle Sequencing Kit, part of the BigDye™ product family, is a complete kit for your PCR and Sequencing needs. This innovative, time-saving kit combines the PCR Purification and Cycle Sequencing Steps, enabling a streamlined protocol and shorter time to results. The kit is available in 24, 100, and 1000-reaction sizes to fit your sequencing throughput needs.

The BigDye™ Direct Cycle Sequencing Kit Gives You These Advantages:
• A streamlined, simple procedure
• Performance of all sequencing steps in a single reaction tube
• Fast migration with excellent resolution starting at the 1st base after the gene-specific primer sequence
• Substantial time savings versus previous workflows

Save Time and Get Better Results with a Unique Streamlined Workflow
The BigDye™ Direct Cycle Sequencing Kits have been developed to minimize complexity and hands-on time. Compared with previous sequencing procedures, which typically require 5 processing steps and take up to 8 hours, Big Dye™ Direct can deliver results in 4 steps taking approximately 5 hours—up to a 40% time savings (Figure 1).

The kit provides reagents for 2 key steps of the sequencing workflow:
• The PCR reaction—using the BigDye™ Direct PCR Master Mix and M13 tailed primer (not provided); PCR is performed in approximately 1 hour.
• The unique one-step PCR purification-cycle sequencing reaction—using the BigDye™ Direct Sequencing Master Mix and the BigDye™ Direct M13 primers provided (M13 tailed primer used for the PCR reaction simplifies the sequencing reaction setup by using 2 sequencing primers independently of the PCR product analyzed).

A One-Tube Protocol Gives You More Confidence
With the BigDye™ Direct Cycle Sequencing Kit, all steps from PCR to loading on the capillary can be performed in the same reaction tube without transferring material from one tube to another. By minimizing the number of manual handling steps, the possibility of experimental variability is reduced; therefore, you can have greater confidence in your data.

An Integrated Solution Ideal for Resequencing
The BigDye™ Direct Cycle Sequencing Kit is the first product designed to maintain conditions leading to high quality data generation throughout the entire read length with faster run times compared to common sequencing workflows. This is achieved through a unique reagent formulation that allows high-resolution data reads closer to the primer and the use of POP-7™ polymer (Figure 2) or SeqStudio cartridge for faster capillary electrophoresis times. The BigDye™ Direct Cycle Sequencing Kit is optimized to work with Applied Biosystems™ Genetic Analyzers and the BigDye XTerminator™ Purification Kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
BigDye™ Direct Cycle Sequencing Kit, 24 rxns, 8-well Strips, 96-well Plate, Fluorescent Dye Terminator Sequencing Technique, 3, Use with 130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, GeneAmp™ 9700, Veriti™ Thermal Cycler
Format
8-well Strip, 96-well Plate
For Use With (Equipment)
3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, SeqStudio Genetic Analyzer, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
For Use With (Application)
Comparative Sequencing (Germline Mutations - 30:70 Heterozygotes), Comparative Sequencing (Germline Mutations - 50:50), GC-Rich Sequencing (>65%), GT-Rich or Difficult Template Sequencing, Standard Sequencing, Gap Closure, Homopolymer A or T, AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations -10:90 Heterozygotes)
No. of Reactions
24
Product Line
BigDye™
Product Type
Direct Cycle Sequencing Kit
Shipping Condition
Dry Ice
Technique
Fluorescent Dye Terminator Sequencing
Template Compatibility
BAC DNA, Cosmids, Lambda DNA, PCR Amplicons (10:90 Heterozygotes), Plasmid DNA (≤15Kb), Genomic DNA (Bacterial), Fosmids, PCR Amplicons (50:50 Heterozygotes), PCR Amplicons (30:70 Heterozygotes)

Contents & Storage

This kit contains BigDye™ Direct PCR Master Mix, Sequencing Master Mix, M13 Fwd Primer, M13 Rev Primer, and Control DNA CEPH 1347-02. Store all components at -15 to -25°C. PCR Master Mix should be stored at 4°C after first use. This kit contains sufficient reagents for 24 reactions.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text