Thermo Scientific Chemicals

Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 445888000
800 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 445888000
800 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 445888000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-41-4
IUPAC Name
sodium methanolate
Molecular Formula
CH3NaO
InChI Key
WQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES
[Na+].C[O-]
Appearance (Form)
Solution
Clarity of solution
Passes test
Titration with HCl
0.48 to 0.52 M
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text