Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals
Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 445901000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametetramethylsilane
Molecular FormulaC4H12Si
InChI KeyCZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILESC[Si](C)(C)C
Molecular Weight (g/mol)88.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Suitability(for 1H-NMR position ref.) Passes test
Residue after evaporation=<0.05 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only