Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 445951000
Price (USD)/ Each
172.65
Online exclusive
192.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
172.65
Online exclusive
192.00 
Save 19.35 (10%)
Add to cart
Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Catalog number445951000
Price (USD)/ Each
172.65
Online exclusive
192.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS23719-80-4
IUPAC Namebromo(cyclopropyl)magnesium
Molecular FormulaC3H5BrMg
InChI KeyQUPLLJGAJVXXBW-UHFFFAOYSA-M
SMILESBr[Mg]C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
Titration Grignard0.40 to 0.60 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only