Methylmagnesium bromide, 1M solution in CPME, AcroSeal™
Methylmagnesium bromide, 1M solution in CPME, AcroSeal™
Methylmagnesium bromide, 1M solution in CPME, AcroSeal™
Methylmagnesium bromide, 1M solution in CPME, AcroSeal™
Thermo Scientific Chemicals

Methylmagnesium bromide, 1M solution in CPME, AcroSeal™

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 446001000
Price (EUR)
58,40
Online Exclusive
68,70
Save 10,30 (15%)
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
58,40
Online Exclusive
68,70
Save 10,30 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Namebromo(methyl)magnesium
Molecular FormulaCH3BrMg
InChI KeyAVFUHBJCUUTGCD-UHFFFAOYSA-M
SMILESC[Mg]Br
Molecular Weight (g/mol)119.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Grignard0.95 to 1.10 M
Appearance (Color)Dark grey
AppearanceMay develop some turbidity or precipitate
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link