Thermo Scientific Chemicals

Methylmagnesium bromide, 1M solution in CPME, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 446008000
800 mL, Each
Thermo Scientific Chemicals

Methylmagnesium bromide, 1M solution in CPME, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 446008000
800 mL, Each
Quantity
Catalog number: 446008000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-16-1
IUPAC Name
bromo(methyl)magnesium
Molecular Formula
CH3BrMg
InChI Key
AVFUHBJCUUTGCD-UHFFFAOYSA-M
SMILES
C[Mg]Br
Appearance (Form)
Solution
Titration Grignard
0.95 to 1.10 M
Appearance (Color)
Dark grey
Appearance
May develop some turbidity or precipitate

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text