Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%

Catalog number: 446085000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%

Catalog number: 446085000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 446085000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10378-23-1
IUPAC Name
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
Molecular Formula
C10H20N2Na4O10
InChI Key
DGWOWDJDIBPYOR-UHFFFAOYSA-N
SMILES
O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Loss on drying
6.4 % to 10.6 %
Titration Complexometric
99.0 to 102.0 % (on dry substance)
Appearance (Form)
Crystalline powder
Solubility
(16% in water) Clear solution
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text