Thermo Scientific Chemicals

Triisopropylsilane, 98%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 446171000
100 mL, Each
Thermo Scientific Chemicals

Triisopropylsilane, 98%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 446171000
100 mL, Each
Quantity
Catalog number: 446171000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6485-79-6
Appearance (Form)
Liquid
Refractive index
1.4330 to 1.4360 (20°C, 589 nm)
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text