N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™, Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 446181000
Price (USD)/ Each
180.65
Online exclusive
212.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
180.65
Online exclusive
212.00 
Save 31.35 (15%)
Add to cart
N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™, Thermo Scientific Chemicals
Catalog number446181000
Price (USD)/ Each
180.65
Online exclusive
212.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(propan-2-yl)({[(propan-2-yl)imino]methylidene})amine
Molecular FormulaC7H14N2
InChI KeyBDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILESCC(C)N=C=NC(C)C
Molecular Weight (g/mol)126.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4320 to 1.4350 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only