1x1 image pixel for data collection
3500 Dx Genetic Analyzer CS2 Install Kit
Product Image
Applied Biosystems™
3500 Dx Genetic Analyzer CS2 Install Kit
This kit includes all of the consumables and accessories needed to perform an instrument qualification—install, operational, or/and performance. This kitRead more
Have Questions?
Catalog NumberQuantity
44620421000 Reactions Kit
Catalog number 4462042
Price (USD)
10,710.00
Each
Add to cart
Quantity:
1000 Reactions Kit
Price (USD)
10,710.00
Each
Add to cart
This kit includes all of the consumables and accessories needed to perform an instrument qualification—install, operational, or/and performance. This kit is specific for the 3500 Dx CS2 instrument and comes with a 50-cm 8-capillary array, POP-6™ polymer, and BigDye® Terminator v1.1 sequencing standard.
For research use only
Specifications
FormatKit
For Use With (Equipment)3500 Dx Genetic Analyzer
For Use With (Application)Sequencing
Product Type3500 Dx Genetic Analyzer CS2 Install Kit
Quantity1000 Reactions Kit
Unit SizeEach
Frequently asked questions (FAQs)
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link