Thermo Scientific Chemicals

3,3-Dimethoxypropanenitrile, 90%, Thermo Scientific Chemicals

Catalog number: 446261000
100 mL, Each
Thermo Scientific Chemicals

3,3-Dimethoxypropanenitrile, 90%, Thermo Scientific Chemicals

Catalog number: 446261000
100 mL, Each
Quantity
Catalog number: 446261000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
57597-62-3
IUPAC Name
3,3-dimethoxypropanenitrile
Molecular Formula
C5H9NO2
InChI Key
JYHSJQNYYLGMEI-UHFFFAOYSA-N
SMILES
COC(CC#N)OC
Refractive index
1.4120 to 1.4160 (20°C, 589 nm)
GC
>=88 %
Appearance (Form)
Liquid
Appearance (Color)
Colorless to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text