Thermo Scientific Chemicals

Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 446290010
1 kg, Each
Thermo Scientific Chemicals

Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 446290010
1 kg, Each
Quantity
Catalog number: 446290010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-72-7
IUPAC Name
sodium cyanosulfanide
Molecular Formula
CNNaS
InChI Key
VGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES
[Na+].[S-]C#N
Titration Argentometric
>=98.0 %
Heavy metals (as Pb)
=<5 ppm
Sulfide (S)
=<0.001 %
Appearance (Form)
Crystalline powder or crystals
Chloride (Cl)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text