Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals
Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 446292500
Price (USD)/ Each
52.65
Online exclusive
57.80 
Save 5.15 (9%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
57.80 
Save 5.15 (9%)
Add to cart
Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals
Catalog number446292500
Price (USD)/ Each
52.65
Online exclusive
57.80 
Save 5.15 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium cyanosulfanide
Molecular FormulaCNNaS
InChI KeyVGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES[Na+].[S-]C#N
Molecular Weight (g/mol)81.07
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.0 %
Heavy metals (as Pb)=<5 ppm
Sulfide (S)=<0.001 %
Appearance (Form)Crystalline powder or crystals
Chloride (Cl)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only