Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals
Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 446301000
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
Add to cart
Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals
Catalog number446301000
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
-
Add to cart
Chemical Identifiers
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfur compounds (as SO4)=<0.003 %
Magnesium (Mg)=<0.001 %
Appearance (Color)Colorless to white
Titration with HCl99.7 to 100.5 % (On dried basis)
Phosphate (PO4)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only