Thermo Scientific Chemicals

Lithium carbonate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 446320010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium carbonate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 446320010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 446320010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
554-13-2
IUPAC Name
dilithium(1+) carbonate
Molecular Formula
CLi2O3
InChI Key
XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]C([O-])=O
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<0.005 %
Sulfate (SO4)
=<0.3 %
Assay
>=98.0 %
Chloride (Cl)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text