Applied Biosystems™

PrepFiler™ BTA Automated Forensic DNA Extraction Kit

Catalog number: 4463354
Have Questions?
Applied Biosystems™

PrepFiler™ BTA Automated Forensic DNA Extraction Kit

Catalog number: 4463354
Have Questions?
Catalog Number
4463354
Unit Size
960 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4463354960 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PrepFiler BTA Automated Forensic DNA Extraction Kit is part of an integrated solution that includes proven PrepFiler BTA reagents for use in automated, high-throughput workflows to provide reliable, simplified, and time-saving DNA purification.

Key features include:
• Improves the overall yield, concentration, and purity of DNA isolated from forensic samples, while enabling removal of PCR inhibitors
• Improves the downstream genotyping success rate to help ensure that critical and often limited casework samples can be successfully analyzed
• Contains specifically developed magnetic particles and reagents that optimize DNA binding and elution of DNA in a small volume
• Offers streamlined protocols to process an extensive variety of forensic sample types

The PrepFiler BTA Automated Forensic DNA Extraction Kit uses magnetic particles with an optimized multi-component surface chemistry to deliver robust and reliable DNA yield from the most challenging forensic samples, including bone, teeth, touch/trace samples, and adhesive substrates. This kit is designed to help improve the overall yield and concentration of DNA recovered, while enabling removal of PCR inhibitors. This means greater quantities of highly purified DNA for improved downstream genotyping results.

To enable customer flexibility in sample processing, plastic consumables for both lysis and purification processing are not included.

Specifications

Bead Type
Magnetic
Sample Type
Forensic Sample
For Use With (Application)
Forensic Testing
High-throughput Compatibility
High-throughput
No. of Reactions
960 Reactions
Product Line
PrepFiler
Shipping Condition
Room Temperature
Target
Forensic DNA

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text