Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 446340010
1 kg, Each
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 446340010
1 kg, Each
Quantity
Catalog number: 446340010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-27-2
IUPAC Name
aluminium(3+) nonahydrate trinitrate
Molecular Formula
AlH18N3O18
InChI Key
SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Titration Complexometric
98.0 to 102.0 %
Appearance (Form)
Crystalline powder or crystals
Chloride (Cl)
=<0.001 %
Magnesium (Mg)
=<0.001 %
Heavy metals (ICP-OES)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text