1x1 image pixel for data collection
Sodium molybdate(VI) dihydrate, ACS reagent
Sodium molybdate(VI) dihydrate, ACS reagent
Sodium molybdate(VI) dihydrate, ACS reagent
Sodium molybdate(VI) dihydrate, ACS reagent
Thermo Scientific Chemicals

Sodium molybdate(VI) dihydrate, ACS reagent

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 446360250
Price (USD)
39.65
Special offer
Online exclusive
Ends: 26-Sep-2025
48.80
Save 9.15 (19%)
Each
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 26-Sep-2025
48.80
Save 9.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS10102-40-6
IUPAC Namedisodium dioxomolybdenumbis(olate) dihydrate
Molecular FormulaH4MoNa2O6
InChI KeyFDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<0.001 %
Titration Complexometric99.5 to 103.0 %
Insoluble matter=<0.005 % (in water)
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.015 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection