Sodium molybdate(VI) dihydrate, ACS reagent, Thermo Scientific Chemicals
Sodium molybdate(VI) dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium molybdate(VI) dihydrate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 446365000
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
Add to cart
Sodium molybdate(VI) dihydrate, ACS reagent, Thermo Scientific Chemicals
Catalog number446365000
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10102-40-6
IUPAC Namedisodium dioxomolybdenumbis(olate)
Molecular FormulaMoNa2O4
InChI KeyTVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<0.001 %
Titration Complexometric99.5 to 103.0 %
Insoluble matter=<0.005 % (in water)
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.015 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only