1x1 image pixel for data collection
Potassium nitrate, 99%, pure, Thermo Scientific Chemicals
Potassium nitrate, 99%, pure, Thermo Scientific Chemicals
Potassium nitrate, 99%, pure, Thermo Scientific Chemicals
Potassium nitrate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium nitrate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 446370100
Price (USD)
-
Request bulk or custom format
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration after Ion exchange>=98.5 %
Appearance (Form)Crystalline powder and/or chunks
Loss on drying=<0.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link