Platinum™ Multiplex PCR Master Mix
Platinum™ Multiplex PCR Master Mix
Applied Biosystems™

Platinum™ Multiplex PCR Master Mix

The Platinum Multiplex PCR Master Mix is designed specifically for endpoint multiplex PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
446426850 Reactions
4464269250 Reactions
44642702000 Reactions
Catalog number 4464268
Price (EUR)
211,00
Each
-
Add to cart
No. of Reactions:
50 Reactions
Request bulk or custom format
Price (EUR)
211,00
Each
Add to cart
Platinum Multiplex PCR Master Mix is a high-specificity, high-throughput solution designed specifically for endpoint multiplex PCR and supports easy multiplexing with minimal optimization.

The performance of the Platinum Multiplex PCR Master Mix over a wide range of amplicon sizes permits the amplification of templates from 50 bp to 2.5 kb, greatly enhancing workflow flexibility. Coupled with its 20-plex capability and absence of primer dimers, it not only provides a high-throughput solution but also boasts high specificity through fewer non-specific primer binding events, resulting in less reaction and primer waste.

Features

  • Amplifies up to 20 amplicons in a single reaction
  • Amplifies products from 50 bp to 2.5 kb
  • Performs multiplex PCR reactions with minimal optimization
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration2X
Fidelity (vs. Taq)1X
Hot StartBuilt-In Hot Start
Multiplex CapabilityUp to 20-plex
No. of Reactions50 Reactions
PolymerasePlatinum Taq DNA Polymerase
Quantity50 Reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
Size (Final Product)2.5 kb or less
Volume1.25 mL
For Use With (Application)Hot-start PCR, Multiplex PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Platinum Multiplex PCR Master Mix, 2X (1 x 1.25 mL)
• GC Enhancer (1 x 0.3 mL)

Store at –15°C to –25°C prior to opening.
Master mix may be stored at –15°C to –25°C or at 4°C (for up to 30 days). The GC Enhancer must be kept at –15°C to –25°C.

Share catalog number, name or link