Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Breakapart | Breakable Strips |
Binding Property | Hydrophilic |
Certified | Yes |
Format | Strip Plate |
Detection Method | Fluorescence |
Array | 8 x 12 |
Packaging | 60 Cases |
Binding Type | Adsorption |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
For Use With (Application) | Fluorescence |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Antibodies (Glycoproteins) |
Unit Size | Case of 60 |
Breakapart | Breakable Strips |
Binding Property | Hydrophobic |
Certified | Yes |
Format | Strip Plate |
Detection Method | Fluorescence |
Array | 8 x 12 |
Packaging | 60 Cases |
Binding Type | Adsorption |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Sterility | Non-sterile |
Surface Treatment | PolySorp |
Well Shape | Round, C-bottom |
For Use With (Application) | Fluorescence |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Lipids |
Unit Size | Case of 60 |
Binding Property | Hydrophobic |
Certified | Yes |
Format | Strip Plate |
Detection Method | Chemiluminescent |
Array | 8 x 12 |
Packaging | 10 Pk., 60 Cases |
Binding Type | Adsorption |
Color | White |
Product Type | Microplate Strip |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | PolySorp |
Well Shape | Round, C-bottom |
For Use With (Application) | Luminescence |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Lipids |
Unit Size | Case of 60 |
Breakapart | Breakable Strips |
Binding Property | Hydrophilic |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Chemiluminescent |
Array | 8 x 12 |
Packaging | 10 Pk., 60 Cases |
Binding Type | Adsorption |
Color | White |
Product Type | Microplate Strip |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
For Use With (Application) | Luminescence |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Antibodies (Glycoproteins) |
Unit Size | Case of 60 |
Catalog Number | Specifications | Unit Size | Surface Treatment | Color | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
446471 | Case of 60 | MaxiSorp | Black | 569.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446473 | Case of 60 | PolySorp | Black | 606.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
463200 | Case of 60 | PolySorp | White | 578.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
463201 | Case of 60 | MaxiSorp | White | Special offer: 413.10 574.00 Save 160.90 (28%) Ends: 30-Jun-2024 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Choice of Surface:
White:
Black:
Note:
Availability may vary by country.