4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%, Thermo Scientific Chemicals
4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 446540010
Price (USD)/ Each
260.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
260.00
Add to cart
4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%, Thermo Scientific Chemicals
Catalog number446540010
Price (USD)/ Each
260.00
-
Add to cart
Chemical Identifiers
CAS16063-80-2
IUPAC Name2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Molecular FormulaC5H5F6NO2
InChI KeyKRNSHCKTGFAMPQ-UHFFFAOYSA-N
SMILESNC(C(C(F)(F)F)C(F)(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or solid
Infrared spectrumConforms
Assay96 to 104 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only