Thermo Scientific Chemicals

Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 446580050
5 g, Each
Thermo Scientific Chemicals

Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 446580050
5 g, Each
Quantity
Catalog number: 446580050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6299-85-0
IUPAC Name
methyl 2,6-dichloropyrimidine-4-carboxylate
Molecular Formula
C6H4Cl2N2O2
InChI Key
DQNQNLWKAGZNIT-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Cl)=NC(Cl)=N1
Appearance (Color)
White to yellow to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
53°C to 59°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text