Thermo Scientific Chemicals

tert-Butyl (3-oxocyclobutyl)carbamate, 95%, Thermo Scientific Chemicals

Catalog number: 446590050
5 g, Each
Thermo Scientific Chemicals

tert-Butyl (3-oxocyclobutyl)carbamate, 95%, Thermo Scientific Chemicals

Catalog number: 446590050
5 g, Each
Quantity
Catalog number: 446590050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
154748-49-9
IUPAC Name
tert-butyl N-(3-oxocyclobutyl)carbamate
Molecular Formula
C9H15NO3
InChI Key
FNHPTFKSPUTESA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC1CC(=O)C1
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
GC
>=94.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text