TaqMan™ Mutation Detection IPC Reagent Kit
TaqMan™ Mutation Detection IPC Reagent Kit
Applied Biosystems™

TaqMan™ Mutation Detection IPC Reagent Kit

Green features
The TaqMan™ Mutation Detection IPC Reagent Kit is a set of optional internal positive control reagents that can be duplexedRead more
Have Questions?
Catalog number 4467538
Price (USD)/ 500 reactions
337.65
Online exclusive
362.00 
Save 24.35 (7%)
-
Add to cart
Price (USD)/ 500 reactions
337.65
Online exclusive
362.00 
Save 24.35 (7%)
Add to cart
TaqMan™ Mutation Detection IPC Reagent Kit
Catalog number4467538
Price (USD)/ 500 reactions
337.65
Online exclusive
362.00 
Save 24.35 (7%)
-
Add to cart
The TaqMan™ Mutation Detection IPC Reagent Kit is a set of optional internal positive control reagents that can be duplexed with any TaqMan™ Mutation Detection assay to provide a positive PCR control result. The IPC reagents can distinguish a mutation target negative result from a PCR failure result. The TaqMan™ Mutation Detection IPC Reagent Kit is preoptimized for use with the TaqMan™ Mutation Detection Assays. The IPC reagents can be duplexed with any TaqMan™ Mutation Detection Assay and gDNA sample, without compromising the mutation detection assay reaction efficiency. This feature allows the mutation detection assays to detect low copies of mutated DNA in the presence of a robust PCR-positive signal from the IPC reagents.

Product Overview

The TaqMan™ Mutation Detection IPC Reagent Kit includes:
50X Exogenous IPC template DNA (200 μL)
A synthetic template that contains the exogenous, non-human DNA target for the assay

10X Exogenous IPC Mix (1 mL)
A primer-limited TaqMan™ assay designed to amplify an exogenous, non-human DNA target; the mix includes:
• Two PCR primers
• A TAMRA™ probe (VIC™ dye-labeled)

Store at –15°C to –25° C, protected from light. Minimize freeze-thaw cycles.

Instrument Compatibility
TaqMan™ Mutation Detection IPC Reagent Kit is compatible with the Applied Biosystems™ ViiA™ 7, 7900HT, 7500, 7500 Fast, and StepOnePlus™ Real-Time PCR Systems.

Sample Type Compatibility
The kit can be used with FFPE samples, cell lines, and fresh frozen samples.
Specifications
Control TemplateNon-human DNA
Detection MethodPrimer-probe
FormFrozen
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Reaction SpeedFast
For Use With (Equipment)7500 Fast System, 7500 System, StepOnePlus™, ViiA™ 7 System
Green FeaturesSustainable packaging
Label or DyeTAMRA, VIC
Product LineTaqMan™
Product TypeMutation Detection Ipc Reagent Kit
Shipping ConditionDry Ice
SpeciesNon-Human
Targetexogeneous, non-human DNA target
For Use With (Application)Genotyping
Concentration10X
Fidelity (vs. Taq)10 X
No. of Reactions500 reactions
Sample TypeFFPE & Fixed Samples, Cells, DNA
Volume200 μL
Unit Size500 reactions
Contents & Storage
Control Primer-Probe Set(s) & Template(s)