Thermo Scientific Chemicals

2,3-Difluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 446770050
5 g, Each
Thermo Scientific Chemicals

2,3-Difluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 446770050
5 g, Each
Quantity
Catalog number: 446770050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
851386-31-7
IUPAC Name
2,3-difluoropyridine-4-carboxylic acid
Molecular Formula
C6H3F2NO2
InChI Key
FFGHMEKFPCGYEG-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(F)C(F)=NC=C1
Titration with NaOH
>=96.0 to =<104.0 %
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text