Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 446792500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
Molecular Weight (g/mol)136.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
pH7.5 to 9.5
Insoluble matter=<0.005 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only