Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 446820025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 446820025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 446820025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
127-08-2
IUPAC Name
potassium acetate
Molecular Formula
C2H3KO2
InChI Key
SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES
[K+].CC([O-])=O
Titration with HClO4
>=99.0 %
Solubility
(5 M in water) Clear colorless solution
Heavy metals (as Pb)
=<5 ppm
pH
6.5 to 9.0
Chloride (Cl)
=<0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text