Magnesium nitrate hexahydrate, for biochemistry
Magnesium nitrate hexahydrate, for biochemistry
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 446830025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13446-18-9
IUPAC Namemagnesium(2+) hexahydrate dinitrate
Molecular FormulaH12MgN2O12
InChI KeyMFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Complexometric98 to 102 %
Insoluble matter=<0.005 %
pH5.0 to 8.2 (at 25°C)
Chloride (Cl)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only