Thermo Scientific Chemicals

Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals

Catalog number: 446882500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals

Catalog number: 446882500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 446882500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-09-3
IUPAC Name
sodium acetate
Molecular Formula
C2H3NaO2
InChI Key
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES
[Na+].CC([O-])=O
Titration with HClO4
>=98.5 %
UV
at 260 nm A: =<0.02
UV
(0.5 M in water)
Loss on drying
=<1 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text