Chemical Identifiers
CAS76-54-0
IUPAC Name2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H10Cl2O5
InChI KeyVFNKZQNIXUFLBC-UHFFFAOYSA-N
SMILESOC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2
View more