Potassium bromate, ACS reagent, Thermo Scientific Chemicals
Potassium bromate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium bromate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 446941000
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
Add to cart
Potassium bromate, ACS reagent, Thermo Scientific Chemicals
Catalog number446941000
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
CAS7758-01-2
IUPAC Namepotassium bromate
Molecular FormulaBrKO3
InChI KeyOCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES[K+].[O-][Br](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH5.0 to 9.0 (5% soln. at 25°C)
Heavy metals (as Pb)=<5 ppm
Sulfate (SO4)=<0.005 %
Titration Iodometric>=99.8 % (Dried basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only