Sodium formate, ACS reagent, Thermo Scientific Chemicals
Sodium formate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium formate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 446951000
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
Add to cart
Sodium formate, ACS reagent, Thermo Scientific Chemicals
Catalog number446951000
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
-
Add to cart
Chemical Identifiers
CAS141-53-7
IUPAC Namesodium formate
Molecular FormulaCHNaO2
InChI KeyHLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES[Na+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO4>=99.0 %
Sulfate (SO4)=<0.001 %
Appearance (Color)White
Insoluble matter=<0.005 %
Iron (Fe)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only