Applied Biosystems™

QuantStudio™ 12K Flex OpenArray™ Gene Expression Starter Kit

Catalog number: 4469604
Have Questions?
Applied Biosystems™

QuantStudio™ 12K Flex OpenArray™ Gene Expression Starter Kit

Catalog number: 4469604
Have Questions?
Catalog Number
4469604
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44696041 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
A variety of starter kits are available to introduce you to the various applications of the QuantStudio™12K Flex OpenArray™ sample block. Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments. The Gene Expression starter kit contains 2 gene expression endogenous control plates. The Gene Expression kit also contains cDNA controls to serve as samples for your experiments. Note that the QuantStudio™12K Flex Software (included with the QuantStudio™ 12K Flex System) offers example experiment files for each application.

The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
384-Well Plate
PCR Method
qPCR
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
QuantStudio™ 12k Flex
Product Line
OpenArray™, QuantStudio™
Product Type
12K Flex Gene Expression Starter Kit
Hot Start
Built-In Hot Start
Sample Type
cDNA

Contents & Storage

• 2X TaqMan™ OpenArray™ Real-Time PCR Master Mix, 1.5 mL
• TaqMan™ OpenArray™ Human Endogenous Control Panels (2 plates)
• Human cDNA controls (brain, liver, lung, and placenta)
• QuantStudio™ 12K Flex OpenArray™ Accessories Starter Kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text