Potassium iodide, 99%, for biochemistry
Potassium iodide, 99%, for biochemistry
Thermo Scientific Chemicals

Potassium iodide, 99%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 447111000
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
Add to cart
Potassium iodide, 99%, for biochemistry
Catalog number447111000
Price (USD)/ Each
106.65
Online exclusive
118.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
Molecular Weight (g/mol)166.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric98.5 to 101.5 %
Insoluble matter=<0.005 % (in water)
Heavy metals (as Pb)=<10 ppm
pH6.0 to 9.0 (5 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only