Applied Biosystems™

Prep-n-Go™ Buffer (for buccal swabs)

Catalog number: 4471406
Have Questions?
Applied Biosystems™

Prep-n-Go™ Buffer (for buccal swabs)

Catalog number: 4471406
Have Questions?
Catalog Number
4471406
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4471406200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Prep-n-Go™ Buffer (for buccal swabs) is a best-in-class buffer designed to enable high-quality direct PCR amplification of single source samples collected on buccal swabs and amplified using the AmpFLSTR™ Identifiler™ Direct PCR Amplification Kit or other Applied Biosystems™ STR direct chemistries. This product expands direct PCR amplification capabilities to laboratories that use a wide variety of substrates, and is readily adaptable to both manual and automated workflows. No purification, extraction, or quantification is required, thus enabling laboratories to significantly expand their throughput capabilities, while also maximizing sample integrity and data quality.

Key Features:

• Designed to provide consistent, interpretable peaks above the detection threshold, and high quality, well-balanced STR profiles
• Helps simplify automation requirements and expand throughput capabilities
• Helps maximize sample integrity and data quality
• Developed according to stringent performance standards to enable higher data reproducibility, reduced artifacts, and increased success rates with Applied Biosystems™ STR direct amplification solutions

Specifications

Buffer
Cell Extraction Buffer
Sample Type
Buccal Swab
For Use With (Application)
PCR
Product Line
Prep-n-Go
Product Type
Buffer
Quantity
1 bottle(s)
Shipping Condition
Room Temperature
Form
Liquid

Contents & Storage

1 Prep-n-Go™ Buffer Bottle (80 mL)
200 tests at 400 μL/test
Store at room temperature and protect from light

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text