Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Thermo Scientific Chemicals

Ammonium phosphate, dibasic, 99%, for biochemistry

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 447142500
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
51.90
Save 10.25 (20%)
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
51.90
Save 10.25 (20%)
Each
Add to cart
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration with HCl>=98.5 %
Solubility(1 M in water) Clear colorless
pH7.5 to 9.0 (1 M in water)
UV(1 M in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link